Product Details
- SNP ID
-
rs3095495
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:56982704 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGGCTGTCTAATCTGGGAGATTTT[G/T]GTGGTTTTCAAACTTTTTGTTCATC
- Phenotype
-
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SCPEP1
PubMed Links
Gene Details
- Gene
- SCPEP1
- Gene Name
- serine carboxypeptidase 1
View Full Product Details