Product Details

SNP ID
rs9471659
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:41794802 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCAAATAAAAGCTAAATATTACA[G/T]AAACCTCACAGTGTTGGAAAAAGCA
Phenotype
MIM: 611389 MIM: 616168
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PRICKLE4 PubMed Links

Gene Details

Gene
PRICKLE4
Gene Name
prickle planar cell polarity protein 4
There are no transcripts associated with this gene.

Gene
TOMM6
Gene Name
translocase of outer mitochondrial membrane 6
There are no transcripts associated with this gene.

Gene
USP49
Gene Name
ubiquitin specific peptidase 49
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286554.1 3900 UTR 3 NP_001273483.1
NM_018561.4 3900 Intron NP_061031.2

View Full Product Details