Product Details

SNP ID
rs7774788
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:28378992 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAATGAATTTATTAATAATACCTA[C/T]GCAAAAAATTTAGGGATGTACAGCA
Phenotype
MIM: 612791 MIM: 603978
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZKSCAN3 PubMed Links

Gene Details

Gene
ZKSCAN3
Gene Name
zinc finger with KRAB and SCAN domains 3
There are no transcripts associated with this gene.

Gene
ZSCAN12
Gene Name
zinc finger and SCAN domain containing 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163391.1 7138 Intron NP_001156863.1
XM_011515014.2 7138 UTR 3 XP_011513316.1
XM_011515015.2 7138 UTR 3 XP_011513317.1
XM_011515017.2 7138 UTR 3 XP_011513319.1
XM_017011528.1 7138 UTR 3 XP_016867017.1

View Full Product Details