Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001893.4 | 2176 | Intron | NP_001884.2 | ||
NM_139062.2 | 2176 | Intron | NP_620693.1 | ||
XM_005256336.3 | 2176 | Intron | XP_005256393.1 | ||
XM_005256337.4 | 2176 | UTR 3 | XP_005256394.1 | ||
XM_017024199.1 | 2176 | Intron | XP_016879688.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001042422.2 | 2176 | UTR 3 | NP_001035887.1 | ||
NM_001042423.2 | 2176 | UTR 3 | NP_001035888.1 | ||
NM_001206950.1 | 2176 | UTR 3 | NP_001193879.1 | ||
NM_001206951.1 | 2176 | UTR 3 | NP_001193880.1 | ||
NM_001206952.1 | 2176 | UTR 3 | NP_001193881.1 | ||
NM_004207.3 | 2176 | UTR 3 | NP_004198.1 | ||
XM_011523607.1 | 2176 | UTR 3 | XP_011521909.1 |