Product Details

SNP ID
rs10410718
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44097560 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAATAATGTTTCAAAGGTTCATGTA[A/T]GTTGTAGCATGAGTCAGTACTTCAT
Phenotype
MIM: 194555
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC100379224 PubMed Links

Gene Details

Gene
LOC100379224
Gene Name
uncharacterized LOC100379224
There are no transcripts associated with this gene.

Gene
ZNF224
Gene Name
zinc finger protein 224
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321645.1 Intron NP_001308574.1
NM_013398.3 Intron NP_037530.2
XM_017027261.1 Intron XP_016882750.1
Gene
ZNF284
Gene Name
zinc finger protein 284
There are no transcripts associated with this gene.

View Full Product Details