Product Details
- SNP ID
-
rs9848567
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:108908441 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTATTTTTACGTTTAGTTGAAAAC[A/G]TCAGGTAATGCCCCAGATGAAATAA
- Phenotype
-
MIM: 605128
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GUCA1C
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77090467] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GUCA1C
- Gene Name
- guanylate cyclase activator 1C
View Full Product Details