Product Details
- SNP ID
-
hCV30475917
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:20141429 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGGGGTGAGTGTGAGTTTGAGGTG[A/T]CTGGGAATGAGTGTGTATGCAAGTG
- Phenotype
-
MIM: 616793
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PLA2G2F
PubMed Links
Gene Details
- Gene
- PLA2G2F
- Gene Name
- phospholipase A2 group IIF
There are no transcripts associated with this gene.
View Full Product Details