Product Details

SNP ID
rs4129183
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:298117 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGTTTCCAGGGAGACGAGGGCGC[A/C]TGCCCGACCCGGGACTTCGTGGTAG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C20orf96 PubMed Links

Gene Details

Gene
C20orf96
Gene Name
chromosome 20 open reading frame 96
There are no transcripts associated with this gene.

Gene
ZCCHC3
Gene Name
zinc finger CCHC-type containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033089.6 555 Silent Mutation GCA,GCC A177A NP_149080.2

View Full Product Details