Product Details

SNP ID
rs9609306
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:31505627 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGTGAGACCCTATCTCTACACA[A/C]AAAAAAAGGCACTTTGGGAGGCTAA
Phenotype
MIM: 612765
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SFI1 PubMed Links

Gene Details

Gene
SFI1
Gene Name
SFI1 centrin binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007467.2 Intron NP_001007468.1
NM_001258325.1 Intron NP_001245254.1
NM_001258326.1 Intron NP_001245255.1
NM_001258327.1 Intron NP_001245256.1
NM_014775.3 Intron NP_055590.2
XM_005261868.2 Intron XP_005261925.1
XM_006724390.2 Intron XP_006724453.1
XM_011530570.2 Intron XP_011528872.1
XM_011530571.2 Intron XP_011528873.1
XM_011530574.2 Intron XP_011528876.1
XM_011530575.2 Intron XP_011528877.1
XM_011530576.2 Intron XP_011528878.1
XM_011530577.2 Intron XP_011528879.1
XM_011530579.2 Intron XP_011528881.1
XM_011530580.2 Intron XP_011528882.1
XM_011530581.2 Intron XP_011528883.1
XM_011530582.2 Intron XP_011528884.1
XM_011530583.2 Intron XP_011528885.1
XM_011530587.2 Intron XP_011528889.1
XM_017029139.1 Intron XP_016884628.1
XM_017029140.1 Intron XP_016884629.1
XM_017029141.1 Intron XP_016884630.1
XM_017029142.1 Intron XP_016884631.1
XM_017029143.1 Intron XP_016884632.1
XM_017029144.1 Intron XP_016884633.1
XM_017029145.1 Intron XP_016884634.1
XM_017029146.1 Intron XP_016884635.1
XM_017029147.1 Intron XP_016884636.1
XM_017029148.1 Intron XP_016884637.1
XM_017029149.1 Intron XP_016884638.1
XM_017029150.1 Intron XP_016884639.1
XM_017029151.1 Intron XP_016884640.1
XM_017029152.1 Intron XP_016884641.1
XM_017029153.1 Intron XP_016884642.1
XM_017029154.1 Intron XP_016884643.1
XM_017029155.1 Intron XP_016884644.1
XM_017029156.1 Intron XP_016884645.1
XM_017029157.1 Intron XP_016884646.1
XM_017029158.1 Intron XP_016884647.1
XM_017029159.1 Intron XP_016884648.1
XM_017029160.1 Intron XP_016884649.1
XM_017029161.1 Intron XP_016884650.1
XM_017029162.1 Intron XP_016884651.1

View Full Product Details