Product Details

SNP ID
rs6527077
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:14888823 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTGTGTTTTTATATCTTTAGATAA[C/T]CTTTTGGGTGTACATCTTTAGCATC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MOSPD2 PubMed Links

Gene Details

Gene
MOSPD2
Gene Name
motile sperm domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177475.1 Intron NP_001170946.1
NM_152581.3 Intron NP_689794.1
XM_005274451.1 Intron XP_005274508.1

View Full Product Details