Product Details

SNP ID
rs7828832
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:38186240 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCTGAAAATACTGGGGCTCTGATC[A/T]CCTTTACCCTTGCTTTTCAAGCCGA
Phenotype
MIM: 603884 MIM: 607281
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
BAG4 PubMed Links

Gene Details

Gene
BAG4
Gene Name
BCL2 associated athanogene 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204878.1 Intron NP_001191807.1
NM_004874.3 Intron NP_004865.1
Gene
LSM1
Gene Name
LSM1 homolog, mRNA degradation associated
There are no transcripts associated with this gene.

View Full Product Details