Product Details

SNP ID
rs12369907
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:76766172 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCTACCGATCTTCAGGTTGCCATT[C/T]GATAGATAGGATATTACTAGAAAGT
Phenotype
MIM: 607799
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZDHHC17 PubMed Links

Gene Details

Gene
ZDHHC17
Gene Name
zinc finger DHHC-type containing 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015336.2 Intron NP_056151.2
XM_005268750.4 Intron XP_005268807.1
XM_011538083.1 Intron XP_011536385.1
XM_017019091.1 Intron XP_016874580.1
XM_017019092.1 Intron XP_016874581.1
XM_017019093.1 Intron XP_016874582.1
XM_017019094.1 Intron XP_016874583.1
XM_017019095.1 Intron XP_016874584.1

View Full Product Details