Product Details

SNP ID
rs8030063
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:64689657 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCAAGACCCCAAGAGCCTTGAATT[C/T]TGGAAAAAAAAAAAAAAAAAGCCAC
Phenotype
MIM: 604152
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OAZ2 PubMed Links

Gene Details

Gene
OAZ2
Gene Name
ornithine decarboxylase antizyme 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301302.1 Intron NP_001288231.1
NM_002537.3 Intron NP_002528.1
Gene
ZNF609
Gene Name
zinc finger protein 609
There are no transcripts associated with this gene.

View Full Product Details