Product Details

SNP ID
rs11591062
Assay Type
Functionally tested
NCBI dbSNP Submissions
60
Location
Chr.1:22434407 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTATTTTCCCAGGTTGTTATTTGC[A/G]TTTAGACTTTATTTATGGATTTTAT
Phenotype
MIM: 612106
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZBTB40 PubMed Links

Gene Details

Gene
ZBTB40
Gene Name
zinc finger and BTB domain containing 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083621.1 Intron NP_001077090.1
NM_014870.3 Intron NP_055685.3
XM_011542499.2 Intron XP_011540801.1
XM_011542500.2 Intron XP_011540802.1
XM_017003003.1 Intron XP_016858492.1

View Full Product Details