Product Details

SNP ID
rs6671231
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:53234845 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATTTTCAAAGGTGGCAAAGTGGT[A/T]AAAAGGAAAGAGTAGCTTTGATGAA
Phenotype
MIM: 602600 MIM: 602603
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC100507564 PubMed Links

Gene Details

Gene
LOC100507564
Gene Name
uncharacterized LOC100507564
There are no transcripts associated with this gene.

Gene
LRP8
Gene Name
LDL receptor related protein 8
There are no transcripts associated with this gene.

Gene
MAGOH
Gene Name
mago homolog, exon junction complex core component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002370.3 Intron NP_002361.1

View Full Product Details