Product Details

SNP ID
rs12853298
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:21710144 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGCTTATAGGCCTCTCTGACTTT[A/G]AGACAGTTTTATAAACATAGAGAAC
Phenotype
MIM: 300226 MIM: 300066
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SMPX PubMed Links

Gene Details

Gene
SMPX
Gene Name
small muscle protein, X-linked
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014332.2 Intron NP_055147.1

View Full Product Details