Product Details

SNP ID
rs11677359
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:54529264 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGTGTTTGAGACCACTTCATCTGG[A/C]CCGAGCTAAAGTCTAGGAAGAAATA
Phenotype
MIM: 182790
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RPL23AP32 PubMed Links

Gene Details

Gene
RPL23AP32
Gene Name
ribosomal protein L23a pseudogene 32
There are no transcripts associated with this gene.

Gene
SPTBN1
Gene Name
spectrin beta, non-erythrocytic 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003128.2 Intron NP_003119.2
NM_178313.2 Intron NP_842565.2
XM_005264517.2 Intron XP_005264574.1
XM_005264518.1 Intron XP_005264575.1
XM_006712087.2 Intron XP_006712150.1
XM_017004779.1 Intron XP_016860268.1
XM_017004780.1 Intron XP_016860269.1
XM_017004781.1 Intron XP_016860270.1

View Full Product Details