Product Details

SNP ID
rs11263842
Assay Type
Functionally tested
NCBI dbSNP Submissions
29
Location
Chr.1:35939348 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTGTGCCAAATTATTTAAAAAGC[A/T]CCAGGTAAAGTAATGACCATGGAGA
Phenotype
MIM: 606228 MIM: 607355
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
AGO1 PubMed Links

Gene Details

Gene
AGO1
Gene Name
argonaute 1, RISC catalytic component
There are no transcripts associated with this gene.

Gene
AGO3
Gene Name
argonaute 3, RISC catalytic component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024852.3 Intron NP_079128.2
NM_177422.2 Intron NP_803171.1
XM_005270575.3 Intron XP_005270632.1
XM_005270576.3 Intron XP_005270633.1
XM_011540879.2 Intron XP_011539181.1
XM_011540882.2 Intron XP_011539184.1
XM_017000523.1 Intron XP_016856012.1
XM_017000524.1 Intron XP_016856013.1
XM_017000525.1 Intron XP_016856014.1
XM_017000526.1 Intron XP_016856015.1
XM_017000527.1 Intron XP_016856016.1
XM_017000528.1 Intron XP_016856017.1

View Full Product Details