Product Details
- SNP ID
-
rs10932243
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:208162930 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTTTTTTTTTTACTACTGAATAAG[A/G]TGACTTTTATGTAGAAATTAGGGAT
- Phenotype
-
MIM: 615536
MIM: 123660
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C2orf80
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs200926418] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C2orf80
- Gene Name
- chromosome 2 open reading frame 80
There are no transcripts associated with this gene.
- Gene
- CRYGA
- Gene Name
- crystallin gamma A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014617.3 |
|
Intron |
|
|
NP_055432.2 |
- Gene
- LOC100507443
- Gene Name
- uncharacterized LOC100507443
There are no transcripts associated with this gene.
View Full Product Details