Product Details

SNP ID
rs12565472
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:211386217 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTAAAAAAAAAGAAAGCTCCAGG[A/C]CGAGATAATTTCACTGGCAAATTTC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LINC00467 PubMed Links

Gene Details

Gene
LINC00467
Gene Name
long intergenic non-protein coding RNA 467
There are no transcripts associated with this gene.

View Full Product Details