Product Details

SNP ID
hCV30874653
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:45645918 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCAATATGACGAAGTCAAATGG[A/T]TGGGATAGAAGCAGATTGTTCTGCC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GPBP1L1 PubMed Links

Gene Details

Gene
GPBP1L1
Gene Name
GC-rich promoter binding protein 1 like 1
There are no transcripts associated with this gene.

Gene
RPS15AP10
Gene Name
ribosomal protein S15a pseudogene 10
There are no transcripts associated with this gene.

View Full Product Details