Product Details

SNP ID
rs11211148
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:45646128 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGGAGCATGGCCTAGTAAGAACC[A/G]GCGTTTCCCTCTCTTTTCGGCATTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPBP1L1 PubMed Links

Gene Details

Gene
GPBP1L1
Gene Name
GC-rich promoter binding protein 1 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021639.4 Intron NP_067652.1
XM_017002019.1 Intron XP_016857508.1
Gene
RPS15AP10
Gene Name
ribosomal protein S15a pseudogene 10
There are no transcripts associated with this gene.

View Full Product Details