Product Details

SNP ID
rs10762275
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:69280863 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGGGAGTGGAATATGATGTGCTAG[A/G]TTCTTTCCAGTTTTCAAATAATATG
Phenotype
MIM: 142600
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HK1 PubMed Links

Gene Details

Gene
HK1
Gene Name
hexokinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000188.2 Intron NP_000179.2
NM_001322364.1 Intron NP_001309293.1
NM_001322365.1 Intron NP_001309294.1
NM_001322366.1 Intron NP_001309295.1
NM_001322367.1 Intron NP_001309296.1
NM_033496.2 Intron NP_277031.1
NM_033497.2 Intron NP_277032.1
NM_033498.2 Intron NP_277033.1
NM_033500.2 Intron NP_277035.2
XM_005269737.1 Intron XP_005269794.1
XM_011539732.1 Intron XP_011538034.1

View Full Product Details