Product Details
- SNP ID
-
rs7672339
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:71006443 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAATTGAAATTGAAAGCAAGGCCAG[A/G]TGTGGTGGTTTATGCATGTAATCCT
- Phenotype
-
MIM: 125450
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DCK
PubMed Links
Gene Details
- Gene
- DCK
- Gene Name
- deoxycytidine kinase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000788.2 |
|
Intron |
|
|
NP_000779.1 |
View Full Product Details