Product Details

SNP ID
rs10772775
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:7815539 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTTGATGTCTAATCTTTCATAGC[A/G]TCAGTAATTACAGTAGTCCCCAGTT
Phenotype
MIM: 611039
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
SLC2A14 PubMed Links

Gene Details

Gene
SLC2A14
Gene Name
solute carrier family 2 member 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286233.1 Intron NP_001273162.1
NM_001286234.1 Intron NP_001273163.1
NM_001286235.1 Intron NP_001273164.1
NM_001286236.1 Intron NP_001273165.1
NM_001286237.1 Intron NP_001273166.1
NM_153449.3 Intron NP_703150.1
XM_005253315.3 Intron XP_005253372.1
XM_005253317.4 Intron XP_005253374.1
XM_011520562.1 Intron XP_011518864.1
XM_011520563.2 Intron XP_011518865.1
XM_011520564.2 Intron XP_011518866.1
XM_011520565.2 Intron XP_011518867.1
XM_017018841.1 Intron XP_016874330.1
XM_017018842.1 Intron XP_016874331.1
XM_017018843.1 Intron XP_016874332.1
XM_017018844.1 Intron XP_016874333.1
XM_017018845.1 Intron XP_016874334.1
XM_017018846.1 Intron XP_016874335.1
XM_017018847.1 Intron XP_016874336.1

View Full Product Details