Product Details

SNP ID
rs12832559
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:8085387 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCTTCACTTCTTAGTTTTCCACA[A/G]CCTTTATACTTACATCAGTTAACTG
Phenotype
MIM: 611623
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NECAP1 PubMed Links

Gene Details

Gene
NECAP1
Gene Name
NECAP endocytosis associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015509.3 Intron NP_056324.2

View Full Product Details