Product Details
- SNP ID
-
rs11065564
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:121414327 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTAGGACAAAGGCAAAAGCCTATTC[A/G]TTCTAAACACTATAGCTTTTATTTT
- Phenotype
-
MIM: 609078
MIM: 608299
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KDM2B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74709796] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KDM2B
- Gene Name
- lysine demethylase 2B
- Gene
- RNF34
- Gene Name
- ring finger protein 34
View Full Product Details