Product Details

SNP ID
hCV30930550
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:18015383 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCAACTGCTTACTGAGTGCTTCA[C/G]TGGCCTTGCCCTGCTCTGGGAAATG
Phenotype
MIM: 608918
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ATPAF2 PubMed Links

Gene Details

Gene
ATPAF2
Gene Name
ATP synthase mitochondrial F1 complex assembly factor 2
There are no transcripts associated with this gene.

Gene
DRC3
Gene Name
dynein regulatory complex subunit 3
There are no transcripts associated with this gene.

View Full Product Details