Product Details

SNP ID
rs13124826
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:214611 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTGCTAGGAAATGCGAATAGCAAG[A/G]ATATACACCCTATGCCACAGTTTCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC107986244 PubMed Links

Gene Details

Gene
LOC107986244
Gene Name
zinc finger protein 141-like
There are no transcripts associated with this gene.

Gene
ZNF876P
Gene Name
zinc finger protein 876, pseudogene
There are no transcripts associated with this gene.

View Full Product Details