Product Details

SNP ID
rs11670041
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35224990 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAGGGCCTTCCTTGCCTCCCGCCA[A/C]TGGGCATCGTTCTCTCTCCACTGAG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM187B PubMed Links

Gene Details

Gene
FAM187B
Gene Name
family with sequence similarity 187 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152481.1 990 Missense Mutation CAG,CAT Q315H NP_689694.1

View Full Product Details