Product Details

SNP ID
hCV31002773
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:143913182 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAATATCATTGTTGATTTGATTCA[A/T]ACTCATTTCATTATCAGTGAAGTTG
Phenotype
MIM: 608497
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR2F1 PubMed Links

Gene Details

Gene
OR2F1
Gene Name
olfactory receptor family 2 subfamily F member 1 (gene/pseudogene)
There are no transcripts associated with this gene.

View Full Product Details