Product Details

SNP ID
rs11169623
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:50938983 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAAGCTTAGCAAGCAATTTTCCTG[A/G]GGTACAAAGAAGTGGCTTCCTAGAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HIGD1C PubMed Links

Gene Details

Gene
HIGD1C
Gene Name
HIG1 hypoxia inducible domain family member 1C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001109619.1 Intron NP_001103089.1
XM_011538649.2 Intron XP_011536951.1
XM_017019783.1 Intron XP_016875272.1
XM_017019784.1 Intron XP_016875273.1
XM_017019785.1 Intron XP_016875274.1
Gene
METTL7A
Gene Name
methyltransferase like 7A
There are no transcripts associated with this gene.

View Full Product Details