Product Details
- SNP ID
-
rs10761323
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:94188863 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTTGATCCAGACAGCTATGTGTAGT[A/G]TATGTCTGAATGCTTTTGACAGATG
- Phenotype
-
MIM: 612145
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIRLET7D
PubMed Links
Gene Details
- Gene
- MIRLET7D
- Gene Name
- microRNA let-7d
There are no transcripts associated with this gene.
- Gene
- MIRLET7DHG
- Gene Name
- MIRLET7D host gene
There are no transcripts associated with this gene.
View Full Product Details