Product Details
- SNP ID
-
rs11129988
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:42914106 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGAGCCTACCCTGGTCCTTGGTTT[G/T]GTGCCAGGAATCATAGCCCGCAGGG
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF662
PubMed Links
Gene Details
- Gene
- ZNF662
- Gene Name
- zinc finger protein 662
View Full Product Details