Product Details

SNP ID
rs13417443
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:241316500 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATAAGCGAGGGGAGAGCGACTAG[A/G]CCCTGTCTGCGGGTACCTTCGGCGA
Phenotype
MIM: 142695 MIM: 601506
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HDLBP PubMed Links

Gene Details

Gene
HDLBP
Gene Name
high density lipoprotein binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243900.2 166 Intron NP_001230829.1
NM_001320965.1 166 Intron NP_001307894.1
NM_001320966.1 166 Intron NP_001307895.1
NM_001320967.1 166 Intron NP_001307896.1
NM_005336.5 166 Intron NP_005327.1
NM_203346.4 166 Intron NP_976221.1
XM_005247002.3 166 Intron XP_005247059.2
XM_005247003.4 166 Intron XP_005247060.2
XM_006712475.3 166 Intron XP_006712538.1
XM_011511058.2 166 Intron XP_011509360.1
XM_011511060.2 166 Intron XP_011509362.1
XM_017003940.1 166 Intron XP_016859429.1
XM_017003941.1 166 Intron XP_016859430.1
XM_017003942.1 166 Intron XP_016859431.1
Gene
SEPT2
Gene Name
septin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008491.2 166 Intron NP_001008491.1
NM_001008492.2 166 Intron NP_001008492.1
NM_001282972.1 166 Silent Mutation AGA,AGG R11R NP_001269901.1
NM_001282973.1 166 UTR 5 NP_001269902.1
NM_001321029.1 166 Silent Mutation AGA,AGG R11R NP_001307958.1
NM_001321030.1 166 Intron NP_001307959.1
NM_001321031.1 166 UTR 5 NP_001307960.1
NM_001321032.1 166 Intron NP_001307961.1
NM_001321033.1 166 UTR 5 NP_001307962.1
NM_001321034.1 166 UTR 5 NP_001307963.1
NM_001321035.1 166 Intron NP_001307964.1
NM_004404.4 166 Intron NP_004395.1
NM_006155.2 166 Intron NP_006146.1
XM_011511237.1 166 Intron XP_011509539.1
XM_011511238.1 166 Intron XP_011509540.1
XM_011511240.1 166 Intron XP_011509542.1
XM_017004189.1 166 Silent Mutation AGA,AGG R11R XP_016859678.1
XM_017004190.1 166 Silent Mutation AGA,AGG R11R XP_016859679.1
XM_017004191.1 166 Silent Mutation AGA,AGG R11R XP_016859680.1
XM_017004192.1 166 Silent Mutation AGA,AGG R11R XP_016859681.1
XM_017004193.1 166 Intron XP_016859682.1
XM_017004194.1 166 UTR 5 XP_016859683.1
XM_017004195.1 166 UTR 5 XP_016859684.1
XM_017004196.1 166 UTR 5 XP_016859685.1
XM_017004197.1 166 Intron XP_016859686.1
XM_017004198.1 166 UTR 5 XP_016859687.1
XM_017004199.1 166 UTR 5 XP_016859688.1
XM_017004200.1 166 Intron XP_016859689.1
XM_017004201.1 166 Intron XP_016859690.1
XM_017004202.1 166 Intron XP_016859691.1
XM_017004203.1 166 Intron XP_016859692.1
XM_017004204.1 166 Intron XP_016859693.1
XM_017004205.1 166 UTR 5 XP_016859694.1
XM_017004206.1 166 Intron XP_016859695.1
XM_017004207.1 166 Intron XP_016859696.1
XM_017004208.1 166 Intron XP_016859697.1
XM_017004209.1 166 Intron XP_016859698.1
XM_017004210.1 166 Intron XP_016859699.1
XM_017004211.1 166 Intron XP_016859700.1
XM_017004212.1 166 Intron XP_016859701.1
XM_017004213.1 166 Silent Mutation AGA,AGG R11R XP_016859702.1

View Full Product Details