Product Details

SNP ID
rs11568634
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.11:62979488 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
ATTCTCCCATTAGGCTCCCACTCAC[C/T]GGATCATTGTGGGATACAGTTCCCC
Phenotype
MIM: 607582
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC22A6 PubMed Links
Additional Information
For this assay, SNP(s) [rs201686570] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
SLC22A6
Gene Name
solute carrier family 22 member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004790.4 1694 Missense Mutation CAG,CGG Q454R NP_004781.2
NM_153276.2 1694 Missense Mutation CAG,CGG Q454R NP_695008.1
NM_153277.2 1694 Missense Mutation CAC,CGC H454R NP_695009.1
NM_153278.2 1694 Missense Mutation CAC,CGC H454R NP_695010.1
XM_017018562.1 1694 Missense Mutation CAG,CGG Q455R XP_016874051.1

View Full Product Details