Product Details

SNP ID
rs7125871
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:87810458 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATCTTTCATTAAGGATAATTATTT[C/T]GAGATTCATCTATGTTGTATGTACC
Phenotype
MIM: 606281
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAB38 PubMed Links

Gene Details

Gene
RAB38
Gene Name
RAB38, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022337.2 Intron NP_071732.1
XM_017017455.1 Intron XP_016872944.1
XM_017017456.1 Intron XP_016872945.1

View Full Product Details