Product Details

SNP ID
rs12564670
Assay Type
Functionally tested
NCBI dbSNP Submissions
25
Location
Chr.1:161683913 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTGAGAAAGGGTTTGCAGGTGATA[C/T]AGTGGAGGCCATAATGCCTCACCTC
Phenotype
MIM: 604590
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FCGR2B PubMed Links

Gene Details

Gene
FCGR2B
Gene Name
Fc fragment of IgG receptor IIb
There are no transcripts associated with this gene.

Gene
RPL31P11
Gene Name
ribosomal protein L31 pseudogene 11
There are no transcripts associated with this gene.

View Full Product Details