Product Details
- SNP ID
-
rs12463916
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:24206836 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GATCGCAGCCTCAGGACAGCGGGGG[C/G]TCCAGGTAGAAGTCACACTGGTTAG
- Phenotype
-
MIM: 604464
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ITSN2
PubMed Links
Gene Details
- Gene
- ITSN2
- Gene Name
- intersectin 2
View Full Product Details