Product Details
- SNP ID
-
rs13122476
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:82486555 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTCTTTCAGACAATACTGTATAGAC[C/T]AGTTTCATTCCTCGTATCACGTCTA
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC105377310
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80327286] are located under a probe and SNP(s) [rs147104155] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC105377310
- Gene Name
- syncytin-1-like
There are no transcripts associated with this gene.
- Gene
- RPS6P6
- Gene Name
- ribosomal protein S6 pseudogene
There are no transcripts associated with this gene.
- Gene
- TMEM150C
- Gene Name
- transmembrane protein 150C
View Full Product Details