Product Details
- SNP ID
-
rs11177823
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:69745893 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGTGTTAGAGTCAGACACACCAGAG[G/T]TTGAATACTAGCTCTGTTACTTTTT
- Phenotype
-
MIM: 608686
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC101928002
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs199779653] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC101928002
- Gene Name
- uncharacterized LOC101928002
There are no transcripts associated with this gene.
- Gene
- RAB3IP
- Gene Name
- RAB3A interacting protein
View Full Product Details