Product Details
- SNP ID
-
rs12724407
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:77698833 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTTTTTAACTCTCCATTTGGGGAT[G/T]ACAAGTCAAAATGTACCACTAATGC
- Phenotype
-
MIM: 615146
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
USP33
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115715356] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- USP33
- Gene Name
- ubiquitin specific peptidase 33
View Full Product Details