Product Details

SNP ID
rs6656763
Assay Type
Functionally tested
NCBI dbSNP Submissions
63
Location
Chr.1:62196120 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTGCATTTTCTTCAATTTCCTCC[A/G]ATTGTTAGGCAAATCGAATTCTTGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
L1TD1 PubMed Links

Gene Details

Gene
L1TD1
Gene Name
LINE1 type transposase domain containing 1
There are no transcripts associated with this gene.

View Full Product Details