Product Details

SNP ID
rs10894152
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:129842124 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAGACCAATAGAAATGATCCAATT[A/C]GAAAAACAGAAAGAAAAATGAAGAC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TMEM45B PubMed Links

Gene Details

Gene
TMEM45B
Gene Name
transmembrane protein 45B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138788.3 Intron NP_620143.1
XM_005271400.4 Intron XP_005271457.1
XM_011542586.2 Intron XP_011540888.1
XM_011542588.2 Intron XP_011540890.1
XM_017017188.1 Intron XP_016872677.1
XM_017017189.1 Intron XP_016872678.1

View Full Product Details