Product Details

SNP ID
rs11195360
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:110874160 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCACCTACCCTCTCTGAATCTGTT[C/T]AGTTGTGTGTAAGGTGGAATACCTA
Phenotype
MIM: 608610
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDCD4 PubMed Links

Gene Details

Gene
PDCD4
Gene Name
programmed cell death 4 (neoplastic transformation inhibitor)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199492.1 Intron NP_001186421.1
NM_014456.4 Intron NP_055271.2
NM_145341.3 Intron NP_663314.1
Gene
PDCD4-AS1
Gene Name
PDCD4 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details