Product Details

SNP ID
rs13306032
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:186672848 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTAAAATTAGCTATAAATAAAATT[A/C]TCTGCTTTTCACTTAAAATAAGTGT
Phenotype
MIM: 600262
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PACERR PubMed Links

Gene Details

Gene
PACERR
Gene Name
PTGS2 antisense NFKB1 complex-mediated expression regulator RNA
There are no transcripts associated with this gene.

Gene
PTGS2
Gene Name
prostaglandin-endoperoxide synthase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000963.3 3457 UTR 3 NP_000954.1

View Full Product Details