Product Details
- SNP ID
-
rs13283731
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:21462594 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAAATAATGTGGTCACTTATGAAC[A/C]CCTTTTAGTATGAGGAGGAAATCTG
- Phenotype
-
MIM: 616356
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MIR31HG
PubMed Links
Gene Details
- Gene
- MIR31HG
- Gene Name
- MIR31 host gene
There are no transcripts associated with this gene.
View Full Product Details