Product Details
- SNP ID
-
rs6650870
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:43870324 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTAGAACATTGGTTAGAAAATATGT[A/T]TGTCGTGCAGTTCAGTTCCAGCTCC
- Phenotype
-
MIM: 614794
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
AGPAT3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113007882] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AGPAT3
- Gene Name
- 1-acylglycerol-3-phosphate O-acyltransferase 3
View Full Product Details