Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001322434.1 | Intron | NP_001309363.1 | |||
NM_001322436.1 | Intron | NP_001309365.1 | |||
NM_001322437.1 | Intron | NP_001309366.1 | |||
NM_001322438.1 | Intron | NP_001309367.1 | |||
NM_001322439.1 | Intron | NP_001309368.1 | |||
NM_001322440.1 | Intron | NP_001309369.1 | |||
NM_001322441.1 | Intron | NP_001309370.1 | |||
NM_001322442.1 | Intron | NP_001309371.1 | |||
NM_001322443.1 | Intron | NP_001309372.1 | |||
NM_001322444.1 | Intron | NP_001309373.1 | |||
NM_001322445.1 | Intron | NP_001309374.1 | |||
NM_001322446.1 | Intron | NP_001309375.1 | |||
NM_001322447.1 | Intron | NP_001309376.1 | |||
NM_001322448.1 | Intron | NP_001309377.1 | |||
NM_001322449.1 | Intron | NP_001309378.1 | |||
NM_001322450.1 | Intron | NP_001309379.1 | |||
NM_001322451.1 | Intron | NP_001309380.1 | |||
NM_001322452.1 | Intron | NP_001309381.1 | |||
NM_001322453.1 | Intron | NP_001309382.1 | |||
NM_012207.2 | Intron | NP_036339.1 | |||
NM_021644.3 | Intron | NP_067676.2 | |||
XM_011539742.1 | Intron | XP_011538044.1 | |||
XM_017016168.1 | Intron | XP_016871657.1 | |||
XM_017016169.1 | Intron | XP_016871658.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001033083.1 | Intron | NP_001028255.1 | |||
NM_022129.3 | Intron | NP_071412.2 | |||
XM_005270028.3 | Intron | XP_005270085.1 | |||
XM_011540060.2 | Intron | XP_011538362.1 | |||
XM_017016513.1 | Intron | XP_016872002.1 | |||
XM_017016514.1 | Intron | XP_016872003.1 |