Product Details

SNP ID
rs7975279
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:24573316 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTAAGGATGGCTCAGAAGGAATAA[A/G]GAAGTGACCTGGATTCTAGAAGTGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00477 PubMed Links

Gene Details

Gene
LINC00477
Gene Name
long intergenic non-protein coding RNA 477
There are no transcripts associated with this gene.

View Full Product Details